Alfa Aesar™ Silver shot, 1-5mm (0.04-0.2 in.), Premion™, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 1218630

Quantity 250g

 View more versions of this product

Catalog No. AA1218630

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-22-4
    MDL Number MFCD00003397
    Chemical Name or Material Silver shot, 1 to 5mm (0.04 to 0.2 in.)
    Grade Premion™

    Specifications

    TSCA TSCA
    MDL Number MFCD00003397
    Odor Characteristic
    Quantity 250g
    Physical Form Shot
    CAS 7440-22-4
    Chemical Name or Material Silver shot, 1 to 5mm (0.04 to 0.2 in.)
    Assay 99.99%, (metals basis)
    Grade Premion™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit