Learn More
Silver, solder alloy, 1.6mm (0.06in) dia, Thermo Scientific Chemicals
For use on copper, stainless, and most other metals
Supplier: Thermo Scientific Chemicals 04103230
Description
Silver, solder alloy is used on copper, stainless, and most other metals. It is used for electronic assemblies, die attachments, thick films. Soldering of jewellery due to the alloys ability to maintain high levels of brightness.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver, solder Alloy | |
Ag:Sn; 3.5:96.5 wt% | |
250 g |
1.6mm (0.06 in.) dia. | |
MFCD00801108 | |
Insoluble in water. |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only