Alfa Aesar™ Silver, solder alloy, 1.6mm (0.06 in.) dia.

For use on copper, stainless, and most other metals

Manufacturer: Alfa Aesar™ 4103236

Quantity 500g

 View more versions of this product

Catalog No. AA4103236

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00801108
    Chemical Name or Material Silver, solder Alloy, 1.6mm (0.06 in.) dia.

    Specifications

    Physical Form Nontoxic, Cadmium and Lead Free
    TSCA TSCA
    MDL Number MFCD00801108
    Avg. Mol. Wt. or Mol. Wt. Range Ag:Sn; 3.5:96.5 wt%
    Quantity 500g
    Chemical Name or Material Silver, solder Alloy, 1.6mm (0.06 in.) dia.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit