Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver sulfate, 99%, extra pure, ACROS Organics™

Click to view available options
Quantity:
10g
100g
500g
Packaging:
Glass bottle
10294-26-5
Ag2O4S
311.79
MFCD00003407
YPNVIBVEFVRZPJ-UHFFFAOYSA-L
silver sulfate,disilver sulfate,disilver 1+ sulfate,unii-8qg6hv4zpo,disilver 1+ sulphate,sulfuric acid, disilver 1+ salt,8qg6hv4zpo,disilver 1+ ion sulfate,sulfuric acid, silver 1+ salt 1:2,sulfuric acid disilver i salt
159865
disilver;sulfate
[O-]S(=O)(=O)[O-].[Ag+].[Ag+]
10294-26-5
Ag2O4S
311.79
MFCD00003407
YPNVIBVEFVRZPJ-UHFFFAOYSA-L
silver sulfate,disilver sulfate,disilver 1+ sulfate,unii-8qg6hv4zpo,disilver 1+ sulphate,sulfuric acid, disilver 1+ salt,8qg6hv4zpo,disilver 1+ ion sulfate,sulfuric acid, silver 1+ salt 1:2,sulfuric acid disilver i salt
159865
disilver;sulfate
[O-]S(=O)(=O)[O-].[Ag+].[Ag+]
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula Ag2SO4
Fieser 01,1015; 03,254; 04,435
Merck Index 15, 8668
Solubility Information Solubility in water: 7.4g/L (20°C). Other solubilities: soluble in nh4oh,nitric acid and sulfuric acid,insoluble in alcohol
Formula Weight 311.79
Physical Form Fine Crystalline Powder
Percent Purity 99%
Grade Extra Pure
Density 1.84g/mL
Insoluble Matter 0.02% max. (in water and AgCl)
Insoluble Matter and AgCl 0.02% max.
Iron (Fe) 10ppm max.
Nitrate 10ppm max.
Packaging Glass bottle
Solubility Solubility in water: 7.4g/L (20°C). Other solubilities: soluble in nh4oh, nitric acid and sulfuric acid, insoluble in alcohol
Substances not ppt by HCl 0.03% max.
Boiling Point 1085.0°C
Color White
Melting Point 652.0°C
Quantity 10g
Chemical Name or Material Silver sulfate
Show More Show Less

GHS H Statement
Causes serious eye damage.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger

EINECSNumber : 233-653-7