Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Silver sulfate, Premion™, 99.99% (metals basis), Ag 68.9% min

Manufacturer:  Alfa Aesar™ 4144322

Quantity 100g

 View more versions of this product

Catalog No. AA4144322


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

10294-26-5
311.8
UN3077
Silver sulfate
Ag2SO4
MFCD00003407
14,8529
Premion™

Specifications

10294-26-5
Ag2SO4
MFCD00003407
14,8529
5.45g/mL
Light Sensitive
White
100g
99.99%, (metals basis)
Powder
311.8
UN3077
Ag 68.9% min.
Odorless
1085°C (dec.)
657°C
Silver sulfate
Premion™

Safety and Handling

SDS & Safety Documents

GHS H Statement
H318
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit