Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver sulfide, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1141622

Quantity 100g

 View more versions of this product

Catalog No. AA1141622


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

21548-73-2
99%
247.8
Silver sulfide
Powder
Ag2S
MFCD00003406

Specifications

21548-73-2
99%
Dark gray to black powder/flake/crystalline/beads
MFCD00003406
Light yellow
Silver sulfide
Powder
Ag2S
247.8
Odorless
100g

Safety and Handling

SDS & Safety Documents

H400-H410: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 244-438-2

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit