Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Silver Thinfoil, 0.0125mm (0.0005 in.) thick, 99.95+% (metals basis)

Click to view available options
Quantity:
25 x 25mm
50 x 50mm
100 x 100mm
7440-22-4
Ag
107.868
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber
23954
CHEBI:9141
silver
[Ag]
7440-22-4
Ag
107.868
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber
23954
CHEBI:9141
silver
[Ag]
Quantity 50 x 50mm
Weight ≈0.08g/25x25mm
Odor Characteristic
Physical Form Foil
Percent Purity ≥99.95%
Chemical Name or Material Silver Thinfoil, 0.0125mm (0.0005 in.) thick, Not certified pinhole free
Assay (metals basis)

EINECSNumber : 231-131-3

TSCA : Yes

Recommended Storage : Ambient temperatures