Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Silver Thinfoil, 0.0125mm (0.0005 in.) thick, 99.95+% (metals basis)
Click to view available options
Quantity:
25 x 25mm
50 x 50mm
100 x 100mm
Specifications
Quantity | 100 x 100mm |
Weight | ≈0.08g/25x25mm |
Odor | Characteristic |
Physical Form | Foil |
Percent Purity | ≥99.95% |
Chemical Name or Material | Silver Thinfoil, 0.0125mm (0.0005 in.) thick, Not certified pinhole free |
Assay | (metals basis) |
Safety and Handling
EINECSNumber : 231-131-3
TSCA : Yes
Recommended Storage : Ambient temperatures