Silver trifluoromethanesulfonate, 98%, Alfa Aesar™

Manufacturer: Alfa Aesar 8872218

Quantity 50g

 View more versions of this product

Catalog No. AA8872218

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2923-28-6
    Physical Form Powder
    Assay Percent Range 98%
    Molecular Formula CAgF3O3S
    Formula Weight 256.93
    MDL Number MFCD00013226
    Synonym Silver triflate
    Chemical Name or Material Silver trifluoromethanesulfonate

    Specifications

    Physical Form Powder
    Formula Weight 256.93
    Quantity 50g
    Color White
    DOT Information DOT Class: 8, Packaging Group: III
    EINECS Number 220-882-2
    Odor Odorless
    Assay Percent Range 98%
    Molecular Formula CAgF3O3S
    MDL Number MFCD00013226
    CAS 2923-28-6
    Synonym Silver triflate
    Health Hazard 1 H314-H318-H400: Causes severe skin burns and eye damage. Causes serious eye damage. Very toxic to aquatic life.
    Chemical Name or Material Silver trifluoromethanesulfonate

    H314-H318-H400: Causes severe skin burns and eye damage. Causes serious eye damage. Very toxic to aquatic life.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit