Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver trifluoromethanesulfonate, 99+%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
2923-28-6
CAgF3O3S
256.92
QRUBYZBWAOOHSV-UHFFFAOYSA-M
silver trifluoromethanesulfonate, silver triflate, agotf, silver trifluoromethanesulphonate, trifluoromethanesulfonic acid silver salt, silver i trifluoromethanesulfonate, silver trifluoromethylsulfonate, methanesulfonic acid, trifluoro-, silver 1+ salt, silver i trifluoromethanesulphonate, silver 1+ ion triflate
76223
silver;trifluoromethanesulfonate
C(F)(F)(F)S(=O)(=O)[O-].[Ag+]
2923-28-6
CAgF3O3S
256.92
QRUBYZBWAOOHSV-UHFFFAOYSA-M
silver trifluoromethanesulfonate, silver triflate, agotf, silver trifluoromethanesulphonate, trifluoromethanesulfonic acid silver salt, silver i trifluoromethanesulfonate, silver trifluoromethylsulfonate, methanesulfonic acid, trifluoro-, silver 1+ salt, silver i trifluoromethanesulphonate, silver 1+ ion triflate
76223
silver;trifluoromethanesulfonate
C(F)(F)(F)S(=O)(=O)[O-].[Ag+]
Elemental Analysis 99.0% min.
Melting Point 356.0°C
Infrared Spectrum Authentic
Quantity 10g
Packaging Glass bottle
Color White to Beige
Assay Percent Range 99+%
Linear Formula CF3SO3Ag
Merck Index 09,302
Solubility Information Solubility in water: soluble.
Formula Weight 256.92
Physical Form Powder
Percent Purity 99+%
Chemical Name or Material Silver trifluoromethanesulfonate
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 220-882-2