Learn More
Silver two part conductive adhesive, EXP. XX/XX Resin and Hardener, Thermo Scientific Chemicals
A conductive epoxy which bonds well to metal, ceramics, plastics, glass and graphite
Supplier: Thermo Scientific Chemicals 1294214
Description
A conductive epoxy which bonds well to metal, ceramics, plastics, glass and graphite
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Limited shelf life, 60 days after shipment
Specifications
Silver two part conductive adhesive | |
EXP. x X/XX Resin and Hardener | |
MFCD00003397 | |
25 g |
Safety and Handling
TSCA : TSCA
storageNote1 :
- Limited shelf life, 60 days after shipment