Alfa Aesar™ Silver two part conductive adhesive, EXP. XX/XX Resin and Hardener

A conductive epoxy which bonds well to metal, ceramics, plastics, glass and graphite

Manufacturer: Alfa Aesar™ 1294203

Quantity 1g

 View more versions of this product

Catalog No. AA1294203

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00003397
    Chemical Name or Material Silver two part conductive adhesive, EXP. x X/XX Resin and Hardener

    Specifications

    MDL Number MFCD00003397
    TSCA TSCA
    Storage Note 1
    • Limited shelf life, 60 days after shipment
    Quantity 1g
    Chemical Name or Material Silver two part conductive adhesive, EXP. x X/XX Resin and Hardener

    • Limited shelf life, 60 days after shipment

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit