Silver wire, 99.95% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 45071G5

Quantity 5m

 View more versions of this product

Catalog No. AA45071G5

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-22-4
    Physical Form Wire, 0.203mm (0.008 in.) dia.
    Assay Percent Range 99.95% (Metals basis)
    MDL Number MFCD00003397
    Chemical Name or Material Silver, Hard

    Specifications

    Quantity 5m
    Color Silver
    EINECS Number 231-131-3
    Physical Form Wire, 0.203mm (0.008 in.) dia.
    Assay Percent Range 99.95% (Metals basis)
    MDL Number MFCD00003397
    CAS 7440-22-4
    Odor Characteristic
    Appearance Silver-colored wire
    Chemical Name or Material Silver, Hard

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit