Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver wire, 99.9985% (metals basis), Alfa Aesar™, Premion™

Click to view available options
Quantity:
100m
25m
5m
7440-22-4
Ag
107.868
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber
23954
CHEBI:9141
silver
[Ag]
7440-22-4
Ag
107.868
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber
23954
CHEBI:9141
silver
[Ag]

Form: approx. 0.51g/m

Quantity 100m
Appearance Silver-colored wire
Assay Percent Range 99.9985% (Metals basis)
Odor Characteristic
Physical Form Wire, ≈0.51g/m
Color Silver
Chemical Name or Material Silver

EINECSNumber : 231-131-3

TSCA : Yes

Recommended Storage : Ambient temperatures