Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SilverXpress™ Silver Staining Kit Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.

Staining is complete in a little over one hour

Manufacturer:  Invitrogen™ LC6100

Catalog No. LC6100


Add to cart

Includes: This kit contains sufficient reagents to stain 25 minigels. Contains: 125mL Sensitizer, 125mL Stainer A, 125mL Stainer B, 125mL Developer, 125mL Stopper solution

Description

Description

The SilverXpress™ Silver Staining Kit is a highly sensitive silver staining kit providing nanogramLevel sensitivity with minimal background. The easy-to-follow protocol requires less total time than standard Coomassie™.

  • Highly sensitive protein staining
  • Crystal clear backgrounds

Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

In-Gel Detection
Room Temperature
Colorimetric
This kit contains sufficient reagents to stain 25 minigels

Contains:
125 mL Sensitizer
125 mL Stainer A
125 mL Stainer B
125 mL Developer
125 Stopper solution

Store kit at 2°C to 8°C. Stable for 6 months when stored under these conditions.
SilverXpress™
Protein
SilverXpress™ Silver Stain
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.