Learn More
Silylating Mixture II according to Horning, MilliporeSigma™ Supelco™
Manufacturer:  MilliporeSigma Supelco 8543210ML
 View more versions of this product
Catalog No. 11-101-8866
Add to cart
Description
Description
Specifications
Specifications
101660-05-3
MFCD00148412
10 mL
4°C
N,O-Bis(trimethylsilyl)acetamide - chlorotrimethylsilane - 1-(trimethylsilyl)imidazole mixture; BSA+TMCS+TMSI 3:2:3; TMSIm/BSA/TMCS 3/3/2 (v/v/v)
For GC derivatization