Learn More
Silylating Mixture II according to Horning, MilliporeSigma™ Supelco™
Manufacturer:  MilliporeSigma Supelco 854321ML
 View more versions of this product
Catalog No. 11-102-0151
Add to cart
Description
Description
Specifications
Specifications
1 mL
101660-05-3
N,O-Bis(trimethylsilyl)acetamide - chlorotrimethylsilane - 1-(trimethylsilyl)imidazole mixture; BSA+TMCS+TMSI 3:2:3; TMSIm/BSA/TMCS 3/3/2 (v/v/v)
4°C
MFCD00148412
For GC derivatization