Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SIM2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568590
Description
This target displays homology in the following species: Cow: 86%; Dog: 93%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Mouse: 91%; Pig: 93%; Rabbit: 92%; Rat: 100%.
This gene represents a homolog of the Drosophila single-minded (sim) gene, which encodes a transcription factor that is a master regulator of neurogenesis. The encoded protein is ubiquitinated by RING-IBR-RING-type E3 ubiquitin ligases, including the parkin RBR E3 ubiquitin protein ligase. This gene maps within the so-called Down syndrome chromosomal region, and is thus thought to contribute to some specific Down syndrome phenotypes. Alternative splicing of this gene results in multiple transcript variants.
Specifications
SIM2 | |
Polyclonal | |
Unconjugated | |
SIM2 | |
BHLHE15; Class E basic helix-loop-helix protein 15; HMC13F06; HMC29C01; mSIM; SIM; SIM bHLH transcription factor 2; SIM transcription factor; sim2; single-minded 2; single-minded family bHLH transcription factor 2; single-minded homolog 2; single-minded homolog 2 (Drosophila); single-minded protein; transcription factor SIM2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
6493 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q14190 | |
SIM2 | |
synthetic peptide directed towards the middle region of human SIM2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction