Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SIM2 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142641
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize all reported isoforms (NP_005060.1; NP_033664.2). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:1,000.
This gene represents a homolog of the Drosophila single-minded (sim) gene, which encodes a transcription factor that is a master regulator of neurogenesis. The encoded protein is ubiquitinated by RING-IBR-RING-type E3 ubiquitin ligases, including the parkin RBR E3 ubiquitin protein ligase. This gene maps within the so-called Down syndrome chromosomal region, and is thus thought to contribute to some specific Down syndrome phenotypes. Alternative splicing of this gene results in multiple transcript variants.
Specifications
SIM2 | |
Polyclonal | |
Unconjugated | |
SIM2 | |
BHLHE15; Class E basic helix-loop-helix protein 15; HMC13F06; HMC29C01; mSIM; SIM; SIM bHLH transcription factor 2; SIM transcription factor; sim2; single-minded 2; single-minded family bHLH transcription factor 2; single-minded homolog 2; single-minded homolog 2 (Drosophila); single-minded protein; transcription factor SIM2 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
20465 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q61079 | |
SIM2 | |
Peptide with sequence C-SDLLYTPSYS. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction