Learn More
Selleck Chemical LLCSupplier Diversity Partner Simeprevir DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S50155MG
Simeprevir is a competitive, reversible, macrocyclic, noncovalent hepatitis C virus (HCV) NS3/4A protease inhibitor that acts directly against the hepatitis C virus. It has a medium inhibitory concentration (IC50) <13 nM for all HCV NS3/4A enzymes(genotypes 1a, 1b, 2, 4, 5, and 6), but has an IC50 value of 37 nM for genotype 3.
Catalog No. 50-194-8438