Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simethicone, USP, 90.5-99%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber Glass Bottle
8050-81-5
C6H18O4Si3
238.46
AMTWCFIAVKBGOD-UHFFFAOYSA-N
3,3,5,5-tetramethyl-2,4-dioxa-3,5-disilahexane; silanedione
O=[Si]=O.CO[Si](C)(C)O[Si](C)(C)C
8050-81-5
C6H18O4Si3
238.46
AMTWCFIAVKBGOD-UHFFFAOYSA-N
3,3,5,5-tetramethyl-2,4-dioxa-3,5-disilahexane; silanedione
O=[Si]=O.CO[Si](C)(C)O[Si](C)(C)C

S1926, 8050-81-5

Spectrum™ Simethicone, USP is an antifoaming agent that is used to reduce bloating from excessive gas. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Quantity 100 mL
CAS Max % 1
Percent Purity 90.5 to 99%
Grade USP