Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Simethicone, USP, 90.5-99%, Spectrum™ Chemical

Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Simethicone, USP is an antifoaming agent that is used to reduce bloating from excessive gas. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
CAS Max % | 1 |
Packaging | Amber Glass Bottle |
Quantity | 500 mL |
Percent Purity | 90.5 to 99% |
Grade | USP |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction