Learn More
BioworldSupplier Diversity Partner Simmons Citrate Agar, 500 g DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 306200731
Simmons Citrate AgarUsed as a differential medium for the identification of enteric bacteria, which utilize citrate as the sole source of carbonph: 6.9Working Concentration: 24.2 g/LDehydrated Appearance: green, homogeneous powderPrepared Appearance: medium to dark green agarSoluble in boiling waterContains:Ammonium Dihydrogen Phosphate Dipotassium Phosphate Sodium Chloride Sodium Citrate Magnesium Sulfate Bromothymol Blue Agar
Catalog No. 50-254-367