Learn More
Essendant Lime Scale Remover, Wintergreen, 1 gal, Bottle, 6/Carton
Supplier:  Essendant SMP50128
Encompass_Preferred
Simple Green Lime Scale Remover uses an organic salt formula to dissolve lime scale, mineral deposits, hard water stains, soap scum and oxidation. The non-abrasive formula won't strip protective finishes, cause discoloration, etch or damage sensitive surfaces like other harsh descalers.
Catalog No. NC9774447
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.