Learn More
Simport™ Scientific M480 Disposable Embedding Cassettes
M480 Disposable Embedding Cassettes for holding and identifying tissue samples in processing, embedding and sectioning procedures during in vitro diagnostics.
$509.00 - $601.00
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
22-040-517
|
Simport™ Scientific
M48012 |
Aqua |
Case of 1500 for $601.00
|
|
|||||
22-040-519
|
Simport™ Scientific
M4807 |
Peach |
Case of 1500 for $601.00
|
|
|||||
22-040-521
|
Simport™ Scientific
M4802 |
White |
Case of 1500 for $509.00
|
|
|||||
Description
- Made of acetal polymer and resistant to chemical action of histological solvents
- Fit securely in microtome chuck adapters
- Accepts standard metal lids that keep specimens safe during processing
- Slanted writing surface for easy sample identification
- Available in three colors
Certifications
CE marked
Specifications
Embedding Cassette |