Simport™ Scientific Polyethylene Flange Plug Caps

Available with two flexible flanges to ensure leakproof seal

Manufacturer: Simport™ Scientific T40410B

 View more versions of this product

Catalog No. 22-038-137

  Check Availability
Add to Cart

    Specifications

    Color Blue
    Product Type Flange Plug Cap
    For Use With (Equipment) 16mm tubes
    Includes Test tubes, round cuvettes, centrifuge tubes, two flexible flanges
    Size (English) 0.62 in.
    Size (Metric) 16mm
    Material Polyethylene

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit