Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Simport™ Scientific T100 BioTube™ Racks
Click to view available options
Product Type:
12-strip Tube Rack
96-plain Tube Rack
Color:
Blue
Red
White
Sterility:
Non-sterile
Sterile
Description
- Made of polypropylene
- Free of RNase, DNase, Pyrogen, and DNA
- Convenient carrying handles on both sides
- Cover, grid plate, and base are keyed to prevent misalignment
- Easy to read ID numbers and letters
- Grid plate stands on 4 legs and can be placed on a lab counter or in a water bath
- These racks and tubes are also ideal for storing, freezing, and transporting reagents and specimens
Specifications
Specifications
Closure Material | Polypropylene |
Color | Red |
Material | Polypropylene |
For Use With | HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting |
Includes | Rack with 96 plain individual tubes |
Quantity | 10/Cs. |
Sterility | Sterile |
Autoclavable | Autoclavable |
Product Type | 96-plain Tube Rack |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction