Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific Cryogenic Tubes

$347.00 - $614.00

Specifications

Certifications/Compliance Certified Rnase-, DNase-, Pyrogen- and DNA-free, CE
Diameter (English) Outer 0.49 in.
Diameter (Metric) Outer 1.25 cm
Autoclavable Autoclavable
Dnase Free Yes
View More Specs

Products 2
Catalog Number Mfr. No. Capacity (Metric) Height (English) Height (Metric) Includes Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Height (English) Height (Metric) Includes Price Quantity & Availability  
10-269-88E
View Documents
Simport™ Scientific
T3104A
4 mL 3.03 in. 7.7 cm Vertical Ribs and Thick Wall, Silicone washer seal and external threads
Case of 1000 for $614.00
Add to cart
 
22-046-103
View Documents
Simport™ Scientific
T3092A
2 mL 1.92 in. 4.9 cm Vertical Ribs and Thick Wall, Lip seal and external threads
Case of 1000 for $347.00
Add to cart
 
Description

Description

Specifications

Specifications

Certified Rnase-, DNase-, Pyrogen- and DNA-free, CE
1.25 cm
Yes
Sterile
Storing biological material, human or animal cells
Cryovial™ Cryogenic Tube
0.49 in.
Autoclavable
Polypropylene
0.25 in. Turn Super Fast External
Yes
Yes
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit