Simport™ Scientific CytoSep™ Funnel Chambers for Hettich Cyto-Systems
Use with Hettich Cyto-Systems to optimize lab throughput with multiple funnel options.
Manufacturer: Simport™ Scientific M9664
 View more versions of this product
Catalog No. 22-045-331
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    For Use With (Application) For cell-rich samples (pleura, ascites, bronchial washings)
    Volume (Metric) 4mL
    For Use With Hettich Cyto-Systems
    Type One-funnel chamber
  • Description & Specifications
Related Products
Product Content Feedback