Simport™ Scientific E-Z LOAD™ Histosette™ II Biopsy Cassette Bases

Manufacturer: Simport™ Scientific M4837BA

 View all options for this product

Catalog No. 05-501-09

  Check Availability
Add to Cart


    Color Peach
    Material High Density Polymer
    Description Biopsy Cassette Base

    • For use with Micro Write™r, Lamb™ and Shur/Mark™ cassette labeling instruments
    • Stacks of 50 cassettes each; case contains two boxes of 20 stacks
    • Bases and covers must be ordered separately

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit