Learn More
Simport Scientific E-Z LOAD Histosette II Biopsy Cassette Bases
For use with Micro Writer, Lamb and Shur/Mark cassette labeling instruments
$426.00
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
05-501-12
|
Simport Scientific
M48310BA |
Lilac |
Case of 2000 for $426.00
|
|
Description
- For use with Micro Writer, Lamb and Shur/Mark cassette labeling instruments
- Stacks of 50 cassettes each; case contains two boxes of 20 stacks
- Bases and covers must be ordered separately
Specifications
Biopsy Cassette Base | |
High Density Polymer |