Learn More
Simport™ Scientific E-Z LOAD™ Histosette™ II Biopsy Cassette Lids
E-Z LOAD™ Histosette™ II Biopsy Cassette Lids for use with Micro Writer™, Lamb™ and SHUR/Mark™ cassette labeling instruments.
$56.51
Specifications
Product Type Biopsy Cassette Cover
View More Specs
Products 1
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
05-501-21
View Documents
Simport™ Scientific
M4838LI
Tan
Case of 2000 for $56.51
Add to cart
Description
Description
  • For use with Micro Writer™, Lamb™ and SHUR/Mark™ cassette labeling instruments
  • Stacks of 50 cassettes each; case contains two boxes of 20 stacks
  • Bases and covers must be ordered separately
Specifications
Specifications
Biopsy Cassette Cover