Simport™ Scientific E-Z LOAD™ Histosette™ II Tissue Cassette Bases

For use with Micro Writer™, Shur/Mark™ and Lamb™ cassette labeling instruments

Manufacturer: Simport™ Scientific M4829BA

 View more versions of this product

Catalog No. 22-038-237

  Check Availability
Add to Cart

    Specifications

    Pore Style Slots
    Material Acetal Polymer
    Description Tissue Cassette
    Writing Face Angle 45°
    Closure Type Integral Lid
    Color Gray

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit