Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Simport Scientific E-Z LOAD Histosette II Tissue Cassette Lids
These cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents
$127.00 - $128.00
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
22-038-254
|
Simport Scientific
M4822LI |
White |
Case of 2000 for $127.00
|
|
22-038-249
|
Simport Scientific
M48210LI |
Lilac |
Case of 2000 for $127.00
|
|
22-038-242
|
Simport Scientific
M4823LI |
Pink |
Case of 2000 for $128.00
|
|
Description
- For use with Micro Writer, Lamb, and Shur/Mark cassette labeling instruments
- Stacks of 50 cassettes each; case contains two boxes of 20 stacks
- Bases and covers must be ordered separately
Specifications
Tissue Cassette Lid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title