Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Simport™ Scientific Graduated Transport Tubes
$141.10 - $364.50
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Polypropylene |
Sterility | Non-sterile |
For Use With (Application) | Storage and transportation of biological materials |
Description
NOTE: Caps are sold separately.Specifications
Autoclavable | |
Polypropylene | |
Non-sterile | |
Storage and transportation of biological materials |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title