Learn More
Simport™ Scientific MultiRack™ Tube Racks
Compact, lightweight and stackable. Simport™ Scientific MultiRack™ Tube Racks are composed of a chemical-resistant acetal polymer that won't shatter or stain.
Manufacturer:  Simport™ Scientific S60016O
 View more versions of this product
Catalog No. 05-407-108
  Check Availability
Add to cart
Specifications
Description
  • Three-tiered racks are stable in water baths
  • Base tier has rounded wells with drain holes
  • Interlocking feet allow stacking
  • With handles for portability
  • Autoclavable
  • Available in 5 colors
Description & Specifications
Specifications
Yes
11.5 x 4.5 x 2.5 in. (29.3 x 11.5 x 6.5cm)
6.5cm
29.3cm
60.0
4.5 in.
Orange
2.5 in.
11.5 in.
Acetal Polymer
Rack, Tube
11.5cm