Simport™ Scientific SLIMSETTE™ M509SL Tissue Processing/Embedding Cassettes in QuickLoad™ Sleeves
A new generation of cassettes, prepackaged for use in automatic cassette labeling instruments
Manufacturer: Simport™ Scientific M50916SL
 View more versions of this product
Catalog No. 23-045-203
  Check Availability
Add to Cart
    Specifications
    Pore Style Slots
    Material Acetal Polymer
    Description Tissue Cassette
    Writing Face Angle 45°
    Closure Type Integral Lid
    Color Fluorescent Pink
  • Description & Specifications
  • Promotions
Related Products
Product Content Feedback