Learn More
Simport™ Scientific SLIMSETTE™ M510 Biopsy Processing/Embedding Cassettes
SLIMSETTE™ M510 Biopsy Processing/Embedding Cassettes are a new generation of cassettes that are more compact and easier to use.
Supplier: Simport™ Scientific M5105
Description
- Specially designed to hold biopsy specimens during the embedding process
- Made of acetal polymer
- Unique recessed cover
- Cassettes are thinner, allowing more to be stacked in automatic labeling machines
- One-piece integral lid
- Large labeling area for easy identification
- Anterior writing area at a 45° angle
Specifications
3 x 500/Cs. | |
Integral Lid | |
Acetal Polymer |
45° | |
Yellow | |
Biopsy Cassette |