Simport™ Scientific SLIMSETTE™ M510SL Biopsy Processing/Embedding Cassettes in Quickload™ Sleeves
A new generation of cassettes, prepackaged for use in automatic cassette labeling instruments
Manufacturer: Simport™ Scientific M5109SL
 View more versions of this product
Catalog No. 23-045-213
  Check Availability
Add to Cart
    Specifications
    Pore Style Slots
    Material Acetal Polymer
    Description Biopsy Cassette
    Writing Face Angle 45°
    Closure Type Integral Lid
    Color Gray
  • Description & Specifications
Related Products
Product Content Feedback