Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Simport™ Scientific SLIMSETTE™ M511 Tissue Processing/Embedding Cassettes
SLIMSETTE™ M511 Tissue Processing/Embedding Cassettes are a new generation of cassettes featuring four square compartments.
Supplier: Simport™ Scientific M5112
Description
- Made of acetal polymer
- Design has a unique recessed cover
- Cassettes are thinner, allowing more to be stacked in automatic labeling machines
- Each cassette has four square compartments measuring 13 mm
- Large labeling area for easy identification
- Anterior writing area at a 45° angle
Specifications
3 x 500/Cs. | |
Integral Lid | |
Acetal Polymer |
45° | |
White | |
Tissue Cassette |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction