Simport™ Scientific SLIMSETTE™ M511 Tissue Processing/Embedding Cassettes
A new generation of cassettes featuring four square compartments
Manufacturer: Simport™ Scientific M5115
 View more versions of this product
Catalog No. 23-045-181
  Check Availability
Add to Cart
    Specifications
    Material Acetal Polymer
    Description Tissue Cassette
    Writing Face Angle 45°
    Closure Type Integral Lid
    Color Yellow
  • Description & Specifications
Related Products
Product Content Feedback