Simport™ Scientific The UniRack™ Jr. Tube Racks

Manufacturer: Simport™ Scientific S50025Y

 View more versions of this product

Catalog No. 22-038-542

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) Cryogenic vials and PCR or microcentrifuge tubes,Cryogenic vials and PCR or microcentrifuge tubes
    Autoclavable No
    Type Rack, Tube
    Holds 0.83mm Tubes
    Positions 25.0,25.0
    Material Polypropylene
    Product Type Rack, Tube
    Color Yellow
    Tube Size 0.83mm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit