Learn More
Simport™ Scientific T100 BIOTUBE™ Tube Racks
T100 BIOTUBE™ Tube Racks are suitable for use in refrigerators, freezers or water baths. Ideal for freezing and storing samples.
Supplier: Simport™ Scientific T1004G
Description
- Stands on four legs and can be removed from the base of the box and placed on a lab counter as a self-standing support
- Can be placed in a refrigerator or freezer shelf for improved air circulation around tubes, or in a water bath to allow controlled warming of the tubes and their contents
- Standard 96-well on center spacing of tubes
- Color coding system using colored interchangeable plastic grids
- Easy to read numbers and letters used on the box cover for sample identification
- Also ideal for storing, freezing and transporting reagents and specimens
- Availaible in red, blue, green and yellow
Specifications
12 Strips of 8 Tubes | |
Green | |
Polypropylene | |
Sterile |
T100 BIOTUBE Tube Rack | |
Tubes (1.2 mL) | |
96 | |
10/Cs. |