Learn More
Simport™ Scientific T100 BIOTUBE™ Tubes
Ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies.
Manufacturer:  Simport™ Scientific T10028
 View more versions of this product
Catalog No. 05-500-20
Add to cart
Description
Description
Ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies.
  • Polypropylene construction
  • Gross volume: 1.2mL, with cap : 1.1mL
  • Caps are made of polyethylene
  • Used for preocedures requiring low surface tension such as protein, nucleic acid work
  • Designed to avoid potentially harmful lubricants while minimizing liquid retention
Applications:
  • Ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting, pharmaceutical quality control, receptor binding assays, RIA and EIA
Specifications
Specifications
Non-sterile
1.2mL
2000xG
T100 Biotube
No
Polyethylene
Ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting, pharmaceutical quality control, receptor binding assays, RIA and EIA
Individual plug caps, 600/Cs.