Sims Portex Hypodermic Needle-Pro™ Device for Injections

Manufacturer: Smiths Medical PM, Inc. 4285

 View more versions of this product

Catalog No. 22-004-239

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Gauge 20 ga
    Length (English) Needle 1 in.
    Length (Metric) Needle 25mm
    Type Hypodermic Needle
    Color Yellow
    For Use With (Application) For intramuscular and subcutaneous injections
    Includes Needle protection device
    Sterile Yes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit