Learn More
Sklar InstrumentsSupplier Diversity Partner Sims Uterine Sound, Sklar, 12-1/2 in. Sims Uterine Sound Non-Graduated Silver Plated. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 905312
Sims Uterine Sound, Sklar, 12-1/2 in. Sims Uterine Sound Non-Graduated Silver Plated. Sklar's Sims Uterine Sound is a malleable, silver plated OB/GYN instrument used for probing and dilating the uterus as well as measuring the length of the cervical canal. It is very similar to the Simpson uterine sound. The instrument is available with or without graduated centimeter markings. This product is straight, but malleable and can be curved as necessary. The handle is a short, smooth thumb grip that features 3 vertical grooves for improved grip. It does not have any measurement markings. Overall, the length is 12-1/2 inches.
Catalog No. 50-143-6806