Learn More
3B Scientific SimShirt System Auscultation Simulator Products for Science Education
When worn by the standardized patient, the SimShirt simulates physiological conditions to test diagnostic and procedural skills. It is controlled wirelessly with easy to use software and responds in real time to diagnosis and treatment with direct feedback.
$8499.00
Specifications
For Use With (Application) To Train and Grow Confidence in Patient Examination.
Product Type Patient Examination Simulator
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
S58108 Products for Science Education
View Documents
3B Scientific
1022827
Large
Each for $8,499.00
Add to cart
S58109 Products for Science Education
View Documents
3B Scientific
1022828
X-Large
Each for $8,499.00
Add to cart
S58110 Products for Science Education
View Documents
3B Scientific
1022829
2 X-Large
Each for $8,499.00
Add to cart
S58111 Products for Science Education
View Documents
3B Scientific
1021552
Medium
Each for $8,499.00
Add to cart
Description
Description
  • The Shirt offers a basic version with all features of the Bionic Hybrid Simulator, except EKG and blood pressure.
  • Includes SimScope Wifi and tablet. Reusable and washable (30°C), available in different sizes
  • Can be worn by students and manikins
  • Simulation of heart, breath, and bowel sounds
  • RFID sensor tags integrated
  • Simulates multiple physiological conditions
  • Real time diagnostics and treatment capabilities
Specifications
Specifications
To Train and Grow Confidence in Patient Examination.
Patient Examination Simulator