Simulaids™ Adult Brad™ CPR Training Manikin

Economical CPR manikin constructed of soft, realistic vinyl plastic over polyurethane foam

Manufacturer: Simulaids, Inc. 2800

 View more versions of this product

Catalog No. 19-819-224

  Check Availability
Add to Cart

    Specifications

    Dimensions (L x W x H) 28 x 18 x 10 in. (71 x 46 x 25cm)
    Height (English) 10 in.
    Height (Metric) 25cm
    Includes 3 disposable/lung airway systems, 3 channel design mouth/nosepieces and kneeling pads
    Material Soft vinyl plastic over polyurethane foam
    Weight (English) 15 lb.
    For Use With (Application) CPR training
    Product Type CPR Training Manikin without Carry Bag

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit