Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simulated Gastric Fluid (without enzyme), J.T.Baker™

Click to view available options
Quantity:
12 x 230.8mL
6 x 961.5mL
Packaging:
Nm Poly Bottle
Poly Bottle

Chemical Identifiers

7647-01-0
ClH
36.458
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
CHEBI:17883
chlorane
Cl
Density Estimated 1 (20°C)
Solubility Information Completely Soluble
Physical Form Liquid
Odor Acrid odor, sharp like hydrochloric acid
Packaging Nm Poly Bottle
Color Colorless
pH 0.4
Quantity 6 x 961.5mL
Chemical Name or Material Simulated Gastric Fluid, without enzyme