Simulated Gastric Fluid (without enzyme), J.T.Baker™

Manufacturer: Avantor Performance Materials D02004

 View more versions of this product

Catalog No. 02-004-274

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7647-01-0
    Chemical Name or Material Simulated Gastric Fluid, without enzyme

    Specifications

    Color Colorless
    pH 0.4
    Quantity 1.9L, 6/Case; NM Poly Bottle
    Physical Form Liquid
    Density Estimated 1 (20°C)
    CAS 7647-01-0
    Solubility Information Completely Soluble
    Odor Acrid odor, sharp like hydrochloric acid
    Chemical Name or Material Simulated Gastric Fluid, without enzyme

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit